Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200268

Sigma-Aldrich

Anti-Tensin 2 (C-terminal) antibody produced in rabbit

enhanced validation

affinity isolated antibody

Synonym(s):

Anti-C1TEN, Anti-TENC1, Anti-TNS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen ~160 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using HEK-293T cells over expressing human tensin 2.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TENC1(23371)

General description

Tensin 2 belongs to the Tensin family. It shares extensive homology with tensin1 at its N- and C-terminals, including the actin-binding domain, the Src Homology 2 (SH2) and c domains. Tensin 2 mRNA is highly expressed in heart, kidney, skeletal muscle and liver.

Immunogen

synthetic peptide corresponding to a sequence located near the C-terminus human tensin 2 (GeneID: 23371), conjugated to KLH. The corresponding sequence is identical in human tensin 2 isoforms 2 and 3, and highly conserved (single amino acid substitution) in mouse tensin 2.

Application

Anti-Tensin 2 (C-terminal) antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Tensin 2 interacts with several binding partners through its Src Homology 2 (SH2)-phosphotyrosine binding (PTB) domains, including Axl receptor tyrosine kinase (RTK), caveolin-1 and with the tumor suppressor gene deleted in liver cancer 1 (DLC1) to regulate DLC1 tumor suppressor activity. Tensin 2 also inhibits Akt signaling and cell survival. It reduces intracellular PtdIns(3,4,5)P3 levels at the plasma membrane. It modulates cytoskeletal dynamics and impacts phosphoinositide-Akt signaling through its PTPase domain. Tensin 2 variant 3 is associated with aggressive tumor behavior in human hepatocellular carcinoma (HCC).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional validation of tensin2 SH2-PTB domain by CRISPR/Cas9-mediated genome editing
Marusugi K, et al.
The Journal of Veterinary Medical Science, 78(9), 1413-1420 (2016)
AXL phosphorylates and up-regulates TNS2 and its implications in IRS-1-associated metabolism in cancer cells
Cheng LC, et al.
Journal of Biomedical Science, 25(1), 80-80 (2018)
Kiyoma Marusugi et al.
The Journal of veterinary medical science, 78(9), 1413-1420 (2016-06-02)
Podocytes are terminally differentiated and highly specialized cells in the glomerulus, and they form a crucial component of the glomerular filtration barrier. The ICGN mouse is a model of glomerular dysfunction that shows gross morphological changes in the podocyte foot
Deleted in liver cancer 1 (DLC1) utilizes a novel binding site for Tensin2 PTB domain interaction and is required for tumor-suppressive function
Chan LK, et al.
PLoS ONE, 4(5), e5572-e5572 (2009)
Tensin2 reduces intracellular phosphatidylinositol 3, 4, 5-trisphosphate levels at the plasma membrane
Hafizi S, et al.
Biochemical and Biophysical Research Communications, 399(3), 396-401 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service