Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200233

Sigma-Aldrich

Anti-VIP36/ LMAN2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GP36B, glycoprotein GP36b, Anti-Lectin, mannose-binding 2, Anti-Vesicular integral protein of 36 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€428.00

€428.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€428.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€428.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~36 kDa

species reactivity

mouse, human, rat

concentration

~1.0 mg/mL

technique(s)

western blot: 2.5-5.0 μg/mL using whole extracts of human HeLa or ACHN cells, rat Rat2 or mouse 3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMAN2(10960)
mouse ... Lman2(66890)
rat ... Lman2(290994)

General description

Anti-VIP36/LMAN2 is produced in rabbit using as immunogen a synthetic peptide of human LMAN2 conjugated to KLH. VIP36 (vesicular-integral protein of 36 kDa) is a member of the leguminous-type (L-type) lectin family, which also includes ERGIC-53, ERGL and VIPL. L-type lectins are type I transmembrane proteins composed of a large luminal domain comprising a carbohydrate recognition domain (CRD) and a short cytosolic domain that mediates sorting within the early secretory pathway. VIP36 localizes mainly to the Golgi apparatus and cycles between the Golgi and the ER.

Application

Anti-VIP36/ LMAN2 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Vesicular-integral protein of 36 kDa (VIP36) binds high mannose type glycoproteins in a Ca2+-dependent manner and may facilitate their sorting, trafficking and quality control.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of the Lectin VIP36 in Post-ER Quality Control of Human alpha1-Antitrypsin
Reiterer V, et al.
Traffic, 11(8), 1044-1055 (2010)
Structural basis for recognition of high mannose type glycoproteins by mammalian transport lectin VIP36
Satoh T, et al.
Test, 282(38), 28246-28255 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service