Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200231

Sigma-Aldrich

Anti-DCAL-1 antibody, Mouse monoclonal

clone UW50, purified from hybridoma cell culture

Synonym(s):

Anti-C-type lectin-like 1, Anti-CLECL1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€525.00

€525.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€525.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€525.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

UW50, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 0.25-0.50 μg/test using 1x106 human Raji cells

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLECL1(160365)

Related Categories

General description

Dendritic cell-associated lectin-1 (DCAL-1), also known as C-type lectin-like-1 (CLECL1), is a novel C-type lectin-like molecule, encoded by the gene mapped to human chromosome 12p13.31. The encoded protein is exclusively expressed in human hematopoietic cells, in particular at high levels in dendritic cells (DCs) and B cells.
Monoclonal Anti-DCAL-1 (mouse IgM isotype) is derived from the hybridoma UW50 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice, immunized with a human DCAL-1 fusion protein

Specificity

Monoclonal Anti-DCAL-1 recognizes human DCAL-1.

Immunogen

human DCAL-1 fusion protein.

Application

Monoclonal Anti-DCAL-1 antibody produced in mouse may be used in flow cytometry.

Biochem/physiol Actions

Dendritic cell-associated lectin-1 (DCAL-1) functions as a T-cell co-stimulatory molecule and increases interleukin-4 secretion. The encoded protein is also implicated in the regulation of the immune response.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20°C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dendritic cell-associated lectin-1: a novel dendritic cell-associated, C-type lectin-like molecule enhances T cell secretion of IL-4.
Ryan EJ
Journal of Immunology, 169, 5638-5648 (2002)
Elizabeth J Ryan et al.
Human immunology, 70(1), 1-5 (2008-11-26)
Dendritic cell-associated lectin-1 (DCAL-1), also known as C-type lectin-like-1 (CLECL1), is a novel C-type lectin-like molecule expressed by antigen presenting cells including dendritic cells (DCs). Here we report that incubation of immature DCs (iDCs) with an anti-DCAL-1 monoclonal antibody (mAb)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service