Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200228

Sigma-Aldrich

Anti-Glycogen Synthase 2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-GYS2, Anti-Glycogen synthase 2 (liver), Anti-LGS

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€503.00

€503.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€503.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€503.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

rat, mouse

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using formalin-fixed paraffin-embedded rat liver
western blot: 1.5-3 μg/mL using mouse liver extracts (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Gys2(232493)

General description

Glycogen synthase 2 (GYS2) codes for a rate-limiting enzyme, glycogen synthase. The encoded protein is exclusively expressed in liver.

Immunogen

synthetic peptide located at the C-terminus of mouse glycogen synthase 2 (GYS2), conjugated to KLH. The corresponding sequence is highly conserved in rat GYS2 (single amino acid substitution) and in human GYS2 (84% identity).

Application

Anti-Glycogen Synthase 2 antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Glycogen synthase 2 (GS-2) plays a vital role as a rate limiting enzyme in glycogenesis pathway in liver and adipose tissue. It acts a target gene of peroxisome proliferator-activated receptors (PPARα, PPARβ/δ and PPARγ), which are transcription factors implicated in regulation of lipid and glucose metabolism. Mutation in the gene is associated with the pathogenesis of inherited monogenic disease glycogen storage disease 0 (GSD0).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycogen synthase 2 is a novel target gene of peroxisome proliferator-activated receptors
Mandard S
Cellular and Molecular Life Sciences, 64, 1145-1157 (2007)
Impaired glucose tolerance and predisposition to the fasted state in liver glycogen synthase knock-out mice.
Irimia JM
The Journal of Biological Chemistry, 285, 12851-12861 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service