Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200042

Sigma-Aldrich

Anti-USP7 antibody, Rat monoclonal

clone USP 3D8, purified from hybridoma cell culture

Synonym(s):

Anti-HAUSP, Anti-TEF1, Monoclonal Anti-USP7 antibody produced in rat

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€390.00

€390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€390.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€390.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

USP 3D8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

human, rat

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 2-4 μg/mL using HeLa cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP7(7874)

General description

Monoclonal Anti-USP7 (rat IgG2a isotype) is derived from the hybridoma USP 3D8 produced by the fusion of mouse myeloma cells (P3X63 AG 8.653) and splenocytes from rat immunized with a peptide.
Ubiquitin specific peptidase 7 (USP7) is a kind of deubiquitylating (DUB) enzyme, which regulates tumor protein p53-mouse double minute 2 homolog (p53-mdm2) pathway. USP7 gene is located on human chromosome 16p13.2.

Specificity

Monoclonal Anti-USP7 recognizes human and rat USP7.

Application

Monoclonal Anti-USP7 antibody produced in rat may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

Overexpression of the USP7 gene results in p53-mediated apoptosis. Ablation of the USP7 gene expression results in p53 accumulation as opposed to the expected destabilization of p53. This effect has been traced to the ability of USP7 to stabilize mouse double minute 2 homolog (MDM2), a ubiquitin ligase that promotes the degradation of p53. Therefore, USP7 appears to play multiple roles in regulating the p53-MDM2 pathway and in maintaining steady-state levels of p53 in the cell. USP7 is considered for treating hematopoietic tumors due to its role in p53 regulation.
Ubiquitin specific peptidase 7 (USP7) promotes oncogenesis and recognizes specific inhibitors. USP7 is overexpressed in hepatocellular carcinoma (HCC) tissues, which contributes to tumor invasion and proliferation. USP7 is important for osteogenic differentiation of human adipose-derived stem cells (hASCs). It is used for hASC based stem cell therapy and bone tissue engineering. USP7 plays complex roles in DNA regulation, p53 stress response and endosomal protein recycling.

Target description

USP7 is ubiquitin specific peptidase 7 (herpes virus-associated). It is associated with induction of apoptosis, interspecies interaction between organisms, protein deubiquitination, regulation of protein stability, and ubiquitin-dependent protein cataboli

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical Significance of Ubiquitin Specific Protease 7 (USP7) in Predicting Prognosis of Hepatocellular Carcinoma and its Functional Mechanisms
Wang Xujing et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 24, 1742-1742 (2018)
Small-molecule inhibitors of USP7 induce apoptosis through oxidative and endoplasmic reticulum stress in cancer cells
Lee G et al.
Biochemical and Biophysical Research Communications, 470(1), 181-186 (2016)
Muyang Li et al.
Molecular cell, 13(6), 879-886 (2004-04-01)
Our previous study showed that ubiquitination of p53 is reversible and that the ubiquitin hydrolase HAUSP can stabilize p53 by deubiquitination. Here, we found that partial reduction of endogenous HAUSP levels by RNAi indeed destabilizes endogenous p53; surprisingly, however, nearly
Use of array CGH to detect exonic copy number variants throughout the genome in autism families detects a novel deletion in TMLHE
Celestino-Soper P, et al.
Human Molecular Genetics, 20(22), 4360-4370 (2011)
Structure of USP7 catalytic domain and three Ubl-domains reveals a connector $\alpha$-helix with regulatory role
Kim R et al.
Journal of Structural Biology, 195(1), 11-18 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service