Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701448

Sigma-Aldrich

Monoclonal Anti-IdU antibody produced in mouse

clone 32D8.D9, purified immunoglobulin

Synonym(s):

Anti-5-iodo-2′-deoxyuridine, Anti-Indoleamine 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€527.00

€527.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€527.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€527.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

32D8.D9, monoclonal

form

buffered aqueous solution

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

Anti-IdU Monoclonal Antibody was purified from ascites fluid by Protein A chromatography. This antibody is specific for IdU. Cross-reactivity is not observed with BrdU, CldU or FdU.

Immunogen

BromodeoxyUridine-KLH followed by hybridoma development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan-Huan Deng et al.
Cell proliferation, 57(5), e13587-e13587 (2023-12-12)
The majority of neocortical projection neurons are generated indirectly from radial glial cells (RGCs) mediated by intermediate progenitor cells (IPCs) in mice. IPCs are thought to be a great breakthrough in the evolutionary expansion of the mammalian neocortex. However, the
Kimberley N Babos et al.
Cell stem cell, 25(4), 486-500 (2019-09-17)
Although cellular reprogramming enables the generation of new cell types for disease modeling and regenerative therapies, reprogramming remains a rare cellular event. By examining reprogramming of fibroblasts into motor neurons and multiple other somatic lineages, we find that epigenetic barriers
DNA polymerase delta interacting protein 3 facilitates the activation and maintenance of DNA damage checkpoint in response to replication stress.
Zhang, et al.
Animal models and experimental medicine, 5, 461-469 (2022)
Wei Huang et al.
Cell, 182(3), 594-608 (2020-07-18)
Human cerebral cortex size and complexity has increased greatly during evolution. While increased progenitor diversity and enhanced proliferative potential play important roles in human neurogenesis and gray matter expansion, the mechanisms of human oligodendrogenesis and white matter expansion remain largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service