Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700447

Sigma-Aldrich

Anti-Swine IgM (μ-chain specific) antibody produced in rabbit

whole antiserum, lyophilized powder

Synonym(s):

Anti-Pig IgM (μ-chain specific) antibody produced in rabbit, Anti-Porcine IgM (μ-chain specific) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€212.00

€212.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
2 ML
€212.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€212.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

swine

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against Swine IgM and Swine Serum. No reaction was observed against Swine IgG.

Immunogen

Swine IgM mu heavy chain

Physical properties

Antibody format: Antiserum

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Reconstitution

Reconstitute with 2.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kara-Lee Aves et al.
Pathogens (Basel, Switzerland), 12(12) (2023-12-22)
F4-positive enterotoxigenic Escherichia coli is associated with diarrhea and poor growth outcomes in neonatal and newly weaned piglets and is thus a major economic and welfare burden in the swine industry. Vaccination of sows with F4 fimbriae protects against the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service