Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700288

Sigma-Aldrich

Anti-goat IgG (H+L)-Biotin antibody produced in donkey

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€208.00

€208.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
2 MG
€208.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€208.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

donkey

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

goat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Goat IgG has two subclasses- IgG1 and IgG2.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Goat IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Biotin, Anti-Donkey Serum, Goat IgG and Goat Serum.

Immunogen

Goat IgG whole molecule

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific IgG for cat allergens in patients with allergic conjunctivitis
Miyama A, et al.
International Ophthalmology (2015)
Ritu Kulshrestha et al.
Iranian journal of basic medical sciences, 23(3), 396-405 (2020-05-23)
The role of N-acetylcysteine (NAC) as an anti-oxidant in attenuating bleomycin-induced pulmonary fibrosis has been reported. However, its effect on parenchymal remodeling via regulating the protease-antiprotease balance is not fully defined. Therefore, the present study was designed to explore the
Tomoya Sasahara et al.
iScience, 24(9), 102936-102936 (2021-08-31)
Amyloid β-protein (Aβ) may contribute to worsening of Alzheimer's disease (AD) through vascular dysfunction, but the molecular mechanism involved is unknown. Using ex vivo blood vessels and primary endothelial cells from human brain microvessels, we show that patient-derived Aβ assemblies, termed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service