Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501265

Sigma-Aldrich

Anti-La Crosse Virus Nucleocapsid antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (La Crosse virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

La Crosse virus ... N(956556)

General description

La Crosse Virus Nucleocapsid Antibody: La Crosse virus (LCV) is a single-stranded RNA virus that can cause encephalitis in humans and is transmitted predominantly by mosquitoes. It is often mistaken for herpes simplex encephalitis. During human infections, LCV initially presents as a nonspecific summertime illness with fever, headache, nausea, vomiting and lethargy. Death occurs in less than 1% of clinical cases.

Immunogen

La Crosse virus nucleocapsid antibody was raised against an 18 amino acid peptide near the center of the La Crosse virus nucleocapsid.The immunogen is located within amino acids 90 - 140 of La Crosse Virus Nucleocapsid.

Application

La Crosse virus nucleocapsid antibody can detect 10 ng La Crosse virus nucleocapsid peptide in ELISA at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service