Skip to Content
Merck
All Photos(3)

Key Documents

SAB3501190

Sigma-Aldrich

Anti-ATR antibody produced in rabbit

purified antibody (Ion exchange)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Check Cart for Availability


Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€370.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody (Ion exchange)

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANTXR1(84168)

Looking for similar products? Visit Product Comparison Guide

General description

ATR Antibody: The Anthrax toxin receptor (ATR) was initially discovered as the tumor endothelial marker 8 (TEM8). This protein, which exists in three isoforms (36, 40, and 60 kDa), is highly expressed in tumor vessels as well as in the vasculature of developing embryos, suggesting that it may normally play a role in angiogenesis. However, it also acts as the receptor for anthrax toxin. Following the binding of this protein by the protective antigen (PA) of anthrax, PA is cleaved and heptamerizes to form the binding site for both edema factor (EF) and lethal factor (LF). This complex is then endocytosed by the cell; acidification in endosomes allows the release of EF and LF into the cytoplasm where they interfere with MAPK signaling and induce apoptosis.

Immunogen

ATR antibody was raised against a peptide corresponding to 13 amino acids near the C-terminus of human ATR.The immunogen is located within the last 50 amino acids of ATR.

Application

ATR antibody can be used for detection of ATR by Western blot at 0.5 to 2 μg/mL. Antibody can also be used for immunocytochemistry starting at 2 μg/mL. For immunofluorescence start at 10 μg/mL.Antibody validated: Western Blot in human samples; Immunocytochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service