Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500978

Sigma-Aldrich

Anti-ACE2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ACEH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€503.00

€503.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€503.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€503.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACE2(59272)

General description

Angiotensin-converting enzyme-2 (ACE2) gene is mapped to human chromosome Xp22.2. The gene codes for a membrane-associated and secreted enzyme. Ace 2 is a human homolog of angiotensin-converting enzyme (ACE) and is found mainly on endothelium. In humans, Ace2 is exclusively expressed in heart, kidney, and testis. The protein is characterized with an N-terminal PD and a C-terminal collectrin-like domain (CLD).

Immunogen

Antibody was raised against a synthetic peptide corresponding to amino acids near the N terminus of human ACE2.

Biochem/physiol Actions

Angiotensin-converting enzyme-2 (ACE2) aids in maturation of angiotensin (Ang) by converting Ang I to Ang1-9. It may also play a role in the local renin-angiotensin system (RAS) in heart and kidney. Ace 2 serves as a functional receptor for severe acute respiratory syndrome–coronavirus (SARS-CoV) and the new coronavirus (SARS-CoV-2), which is responsible for causing coronavirus disease 2019 (COVID-19). Reduced expression of Ace 2 might cause cardiovascular diseases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500978.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaśmina Bałaban et al.
Nanotechnology, science and applications, 16, 1-18 (2023-01-27)
The experiments aimed to document the presence of the ACE2 receptor on human muscle cells and the effects of the interaction of these cells with the spike protein of the SARS-CoV-2 virus in terms of induction of pro-inflammatory proteins, as
Emily J Sherman et al.
Scientific reports, 11(1), 15900-15900 (2021-08-07)
The membrane protein angiotensin-converting enzyme 2 (ACE2) is a physiologic regulator of the renin-angiotensin system and the cellular receptor for the SARS-CoV-2 virus. Prior studies of ACE2 expression have primarily focused on mRNA abundance, with investigation at the protein level
M Donoghue et al.
Circulation research, 87(5), E1-E9 (2000-09-02)
ACE2, the first known human homologue of angiotensin-converting enzyme (ACE), was identified from 5' sequencing of a human heart failure ventricle cDNA library. ACE2 has an apparent signal peptide, a single metalloprotease active site, and a transmembrane domain. The metalloprotease
Bina Lee et al.
Cell reports. Medicine, 5(5), 101570-101570 (2024-05-16)
While an association between Parkinson's disease (PD) and viral infections has been recognized, the impact of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) on PD progression remains unclear. Here, we demonstrate that SARS-CoV-2 infection heightens the risk of PD using
ACE2 is on the X chromosome: could this explain COVID-19 gender differences?
Esther Culebras et al.
European heart journal (2020-06-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service