Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500661

Sigma-Aldrich

Anti-VKORC1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

mol wt

18 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VKORC1(79001)

General description

Vitamin K epoxide reductase complex subunit 1 (VKORC1), also known as vitamin K clotting factor deficiency type 2 (VKCFD2), is an integral, 163-amino acid long transmembrane protein. It is encoded by the gene mapped to a 20Mb region on human chromosome 16p12–q21. VKORC1 is widely expressed in various tissues and organs.

Immunogen

Antibody was raised against a 15 amino acid peptide near the amino terminus of human VKORC1.

Biochem/physiol Actions

Vitamin K epoxide reductase complex subunit 1 (VKORC1) enzyme plays a vital role in vitamin K cycle. It catalyzes the conversion of inactive vitamin K to vitamin K hydroquinone, which is crucial for the post-translational γ-carboxylation of vitamin K–dependent clotting factors II (prothrombin), VII, IX, and X. VKORC1 acts as a key target for oral anticoagulants and is also involved in drug metabolism.{4} Genetic variation in the gene leads to warfarin resistance and multiple coagulation factor deficiency type 2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500661.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High prevalence of VKORC1*3 (G9041A) genetic polymorphism in north Indians: A study on patients with cardiac disorders on acenocoumarol.
Sehgal T
Drug Discoveries & Therapeutics (2015)
G-1639A but Not C1173T VKORC1 Gene Polymorphism Is Related to Ischemic Stroke and Its Various Risk Factors in Ukrainian Population.
Dubovyk YI
BioMed Research International (2016)
VKCFD2 - from clinical phenotype to molecular mechanism.
Czogalla KJ
Hamostaseologie (2016)
Mutations in VKORC1 cause warfarin resistance and multiple coagulation factor deficiency type 2.
Rost S
Nature (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service