Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500544

Sigma-Aldrich

Anti-NOD3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CLR16.2, Anti-NLRC3, Anti-Nucleotide-binding oligomerization domain containing 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€360.00

€360.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1467


Select a Size

Change View
100 μG
€360.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€360.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1467

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRC3(197358)

General description

NLR family CARD domain containing 3 (NLRC3), also referred to as nucleotide-binding oligomerization domain containing 3 (NOD3), is encoded by the gene mapped to human chromosome 16p13.3. The encoded protein belongs to the NLR protein family and is abundantly expressed in purified T cells, B cells and natural killer (NK) cells, but also in thymus, uterus and kidney.

Immunogen

NOD3 antibody was raised against an 18 amino acid peptide near the center of human NOD3.

Biochem/physiol Actions

NLR family CARD domain containing 3 (NLRC3) negatively regulates nuclear factor-κB (NF-κB) activity and innate immune signaling induced by the DNA sensor STING (stimulator of interferon gene). NLRC3 inhibits mammalian target of rapamycin (mTOR) pathways involved in providing protection against colorectal cancer. Thus, downregulated expression of the gene has been observed in colorectal cancer and Wegener′s granulomatosis patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500544.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression profile of innate immune receptors, NLRs and AIM2, in human colorectal cancer: correlation with cancer stages and inflammasome components
Liu R, et al.
Oncotarget, 6(32), 33456-33456 (2015)
NLRC3, a member of the NLR family of proteins, is a negative regulator of innate immune signaling induced by the DNA sensor STING
Zhang L, et al.
Immunity, 40(3), 329-341 (2014)
NLR proteins: integral members of innate immunity and mediators of inflammatory diseases
Wilmanski, et al.
Journal of Leukocyte Biology, 83(1), 13-30 (2008)
NLRC3 is an inhibitory sensor of PI3K?mTOR pathways in cancer
Karki R, et al.
Nature, 540(7634), 583-583 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service