Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500458

Sigma-Aldrich

Anti-PLEKHM1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AP162, Anti-B2, Anti-OPTB6, Anti-Pleckstrin homolog domain containing family M member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€361.00

€361.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€361.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€361.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLEKHM1(9842)

General description

The gene PLEKHM1 (pleckstrin homology and RUN domain containing M1) is mapped to human chromosome 17q21.31. The encoded protein is widely expressed. The protein localizes in the cytoplasm and late endosomal/lysosomal compartments. PLEKHM1 has a RH (rubicon homologous) domain, two PH (pleckstrin homology) domains and a LIR (LC3 interaction region).[1]

Immunogen

PLEKHM1 antibody was raised against an 18 amino acid peptide from near the amino terminus of human PLEKHM1.

Biochem/physiol Actions

PLEKHM1 (pleckstrin homology and RUN domain containing M1) is involved in the end stages of endocytic and autophagy pathways. Here, it interacts with RAB7 (member of RAS oncogene family), the HOPS (homotypic fusion and vacuole protein sorting) complex and ATG8 (autophagy-related protein 8) proteins. It plays an important role in autophagosome-lysosome fusion and endosome and autophagosome maturation. PLEKHM1 also participates in osteoclast function and bone resorption. In presence of Salmonella enterica Typhimurium, the PLEKHM1-RAB7-HOPS complex targets the Salmonella effector protein SifA (Salmonella-induced filament protein A) and controls vacuoles consisting of Salmonella.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500458.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David G McEwan et al.
Molecular cell, 57(1), 39-54 (2014-12-17)
The lysosome is the final destination for degradation of endocytic cargo, plasma membrane constituents, and intracellular components sequestered by macroautophagy. Fusion of endosomes and autophagosomes with the lysosome depends on the GTPase Rab7 and the homotypic fusion and protein sorting
Simone de Jong et al.
BMC genomics, 13, 458-458 (2012-09-07)
Chromosome 17q21.31 contains a common inversion polymorphism of approximately 900 kb in populations with European ancestry. Two divergent MAPT haplotypes, H1 and H2 are described with distinct linkage disequilibrium patterns across the region reflecting the inversion status at this locus. The
Liesbeth Van Wesenbeeck et al.
The Journal of clinical investigation, 117(4), 919-930 (2007-04-04)
This study illustrates that Plekhm1 is an essential protein for bone resorption, as loss-of-function mutations were found to underlie the osteopetrotic phenotype of the incisors absent rat as well as an intermediate type of human osteopetrosis. Electron and confocal microscopic
Keisuke Tabata et al.
Molecular biology of the cell, 21(23), 4162-4172 (2010-10-15)
The endocytic and autophagic pathways are involved in the membrane trafficking of exogenous and endogenous materials to lysosomes. However, the mechanisms that regulate these pathways are largely unknown. We previously reported that Rubicon, a Beclin 1-binding protein, negatively regulates both
David G McEwan et al.
Autophagy, 11(4), 720-722 (2015-04-24)
The endosomal system and autophagy are 2 intertwined pathways that share a number of common protein factors as well as a final destination, the lysosome. Identification of adaptor platforms that can link both pathways are of particular importance, as they

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service