Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500449

Sigma-Aldrich

Anti-LIMP2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD36L2, Anti-Lysosomal integral membrane protein 2, Anti-SCARB2, Anti-SR-B2, Anti-Scavenger receptor class B member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€481.00

€481.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€481.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€481.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 53 kDa

species reactivity

mouse, rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCARB2(950)

General description

Lysosomal integral membrane protein type 2 (LIMP2) is an N-glycosylated type III transmembrane protein. It is composed of a lumenal domain, two transmembrane domains, and a cytoplasmic domain. LIMP2 belongs to the CD36 family of scavenger receptor proteins and is found abundantly in the lysosome. The LIMP2 gene is located on the human chromosome at 4q21.1.

Immunogen

LIMP2 antibody was raised against a 16 amino acid peptide from near the center of human LIMP2.

Application

Anti-LIMP2 antibody produced in rabbit has been used in immunofluorescence (1:100) and western blotting.[1]

Biochem/physiol Actions

Lysosomal integral membrane protein type 2 (LIMP2) binds to β-glucocerebrosidase. Mutations in the LIMP 2 gene are associated with action myoclonus-renal failure syndrome, a fatal autosomal recessive disorder. Higher levels of LIMP2 lead to enlargement of early or late endosomes/lysosomes. Overexpression of LIMP2 is associated with impairment of endocytotic membrane trafficking.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500449.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wo-Tu Tian et al.
Seizure, 57, 80-86 (2018-04-02)
To describe the clinical and genetic features of a Chinese progressive myoclonus epilepsy (PME) patient related with SCARB2 mutation without renal impairment and review 27 SCARB2-related PME patients from 11 countries. The patient was a 27-year-old man with progressive action
Judith Blanz et al.
Human molecular genetics, 19(4), 563-572 (2009-11-26)
Action myoclonus-renal failure syndrome (AMRF) is caused by mutations in the lysosomal integral membrane protein type 2 (LIMP-2/SCARB2). LIMP-2 was identified as a sorting receptor for beta-glucocerebrosidase (beta-GC), which is defective in Gaucher disease. To date, six AMRF-causing mutations have
Abolfazl Yari et al.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology (2021-03-28)
Progressive myoclonic epilepsy-4 with or without renal failure (EPM4) is a rare neurological autosomal recessive disorder caused by mutations in SCARB2 gene. In this study, we described clinical features and genetic causes of an Iranian family with two affected individuals
LIMP-2 is a receptor for lysosomal mannose-6-phosphate-independent targeting of beta-glucocerebrosidase.
Reczek, et al.
Cell, 131, 770-783 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service