Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500152

Sigma-Aldrich

Anti-NSA1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NOP seven-associated protein 1, Anti-WD repeat-containing protein 74, Anti-WDR74

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€361.00

€361.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€361.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€361.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDR74(54663)

General description

WD repeat domain 74 (WDR74), also known as NOP seven-associated protein 1 (NSA1), is encoded by the gene mapped to human chromosome 11q12.3. The encoded protein is mainly localized in the nucleus and it contains six WD40 repeats.

Immunogen

NSA1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human NSA1.

Biochem/physiol Actions

WD repeat domain 74 (WDR74) along with the nucleolar AAA-ATPase NVL2, plays a vital role in an early cleavage of the pre-rRNA processing pathway. It is also implicated in various biological processes, including cell cycle control and apoptosis. In mouse, this protein plays an essential role in RNA transcription, processing and/or stability during preimplantation development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500152.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide analysis of non-coding regulatory mutations in cancer
Weinhold N, et al.
Nature Genetics, 46(11), 1160-1160 (2014)
Genes Involved in Human Ribosome Biogenesis are Transcriptionally Upregulated in Colorectal Cancer
Mansilla F, et al.
Scholarly Research Exchange (2009)
Wdr74 Is Required for Blastocyst Formation in the Mouse
Maserati M, et al.
PLoS ONE, 6(7), e22516-e22516 (2011)
WDR74 participates in an early cleavage of the pre-rRNA processing pathway in cooperation with the nucleolar AAA-ATPase NVL2.
Hiraishi N, et al.
Biochemical and Biophysical Research Communications, 495(1), 116-123 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service