SAB3500120
Monoclonal Anti-ORAI3 antibody produced in mouse
clone 2H2G9, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-Calcium release-activated calcium channel protein 3, Anti-TMEM142C, Anti-Transmembrane protein 142C
Select a Size
€371.00
Select a Size
About This Item
€371.00
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2H2G9, monoclonal
form
buffered aqueous solution
mol wt
predicted mol wt 32 kDa
species reactivity
rat, human
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Immunogen
Features and Benefits
Target description
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service