Skip to Content
Merck
All Photos(2)

Key Documents

SAB2900881

Sigma-Aldrich

Anti-NLRC3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CARD15-like protein, Caterpiller 16.2, Caterpiller protein 16.2, NLRC3, NOD-like receptor C3, NOD3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€569.00

€569.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1467


Select a Size

Change View
50 μG
€569.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€569.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1467

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry: 5 μg/mL
western blot: 1-2 μg/mL (Recombinant Protein)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRC3(197358)

Immunogen

An 18 amino acid peptide near the center of human NOD3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Yu Ma et al.
Human pathology, 82, 273-281 (2018-08-07)
NLRC3 is a member of the nucleotide-binding domain and leucine-rich repeat (NLR) family protein that plays a role in inflammation and immunity. Although chronic inflammation has been identified as a hallmark of cancer, NLRC3 expression correlation with the development and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service