Skip to Content
Merck
All Photos(4)

Key Documents

SAB2702245

Sigma-Aldrich

Monoclonal Anti-Dengue virus NS3 protein antibody produced in mouse

clone GT2811, affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€446.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT2811, monoclonal

form

buffered aqueous solution

mol wt

69kDa

species reactivity

Dengue virus

concentration

1mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1000-10000

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Dengue virus NS3 protein.

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:2000, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayan Modak et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 37(8), e23062-e23062 (2023-06-30)
The factors that drive dengue virus (DENV) evolution, and selection of virulent variants are yet not clear. Higher environmental temperature shortens DENV extrinsic incubation period in mosquitoes, increases human transmission, and plays a critical role in outbreak dynamics. In the
Ayan Modak et al.
IUBMB life, 76(5), 267-285 (2023-11-30)
Extensive vascular leakage and shock is a major cause of dengue-associated mortality. At present, there are no specific treatments available. Sphingolipid pathway is a key player in the endothelial barrier integrity; and is mediated through the five sphingosine-1-phosphate receptors (S1PR1-S1PR5).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service