Skip to Content
Merck
All Photos(1)

Key Documents

SAB2700244

Sigma-Aldrich

Anti-CD68 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CD68, DKFZp686M18236, GP110, SCARD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:500-1:3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD68(968)

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 194 of CD68 according to NP_001242

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes a 110-kD transmembrane glycoprotein that is highly expressed by human monocytes and tissue macrophages. It is a member of the lysosomal/endosomal-associated membrane glycoprotein (LAMP) family. The protein primarily localizes to lysosomes and endosomes with a smaller fraction circulating to the cell surface. It is a type I integral membrane protein with a heavily glycosylated extracellular domain and binds to tissue- and organ-specific lectins or selectins. The protein is also a member of the scavenger receptor family. Scavenger receptors typically function to clear cellular debris, promote phagocytosis, and mediate the recruitment and activation of macrophages. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shogo Kumagai et al.
Lung cancer (Amsterdam, Netherlands), 85(3), 457-464 (2014-07-22)
Increasing evidence suggests that an elevated peripheral monocyte count at presentation predicts a poor prognosis in various types of malignancy, including malignant lymphoma. In lung adenocarcinoma, tumor-associated macrophages (TAMs) were reported to be associated with a poor prognosis. However, it
Kalliopi Pitarokoili et al.
Journal of neuroimmunology, 274(1-2), 38-45 (2014-07-10)
Laquinimod is an immunomodulatory drug with neuroprotective potential. We used the animal model of experimental autoimmune neuritis (EAN) in the Lewis rat to study the effects of laquinimod treatment. After immunization with the neuritogenic peptide aa 53-78 of P2 myelin
Yunmei Liao et al.
Laboratory investigation; a journal of technical methods and pathology, 94(7), 706-715 (2014-05-28)
Tumor-associated macrophages are a prominent component of lung cancer stroma and contribute to tumor progression. The protein V-set and Ig domain-containing 4 (VSIG4), a novel B7 family-related macrophage protein that has the capacity to inhibit T-cell activation, has a potential
Manabu Shigeoka et al.
Cancer science, 104(8), 1112-1119 (2013-05-08)
Tumor associated macrophages (TAMs) are the most abundant cancer stromal cells educated by tumor microenvironment to acquire trophic functions facilitating angiogenesis, matrix breakdown and cancer cell motility. Tumor associated macrophages have anti-inflammatory properties or "alternatively" activated (M2) phenotype expressing CD204
Cristina Gonçalves et al.
World journal of gastroenterology, 20(42), 15780-15786 (2014-11-18)
To characterize clinical, laboratorial, and histological profile of pediatric autoimmune gastritis in the setting of unexplained iron deficiency anemia investigation. A descriptive, observational study including pediatric patients with a diagnosis of autoimmune gastritis (positive parietal cell antibody and gastric corpus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service