Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501722

Sigma-Aldrich

Anti-TPO antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

MSA, TDH2A, TPX, thyroid peroxidase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€477.00

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPO(7173)

General description

Thyroid peroxidase (TPO) is a membrane-bound, glycosylated, heme-containing peroxidase. It is encoded by the gene mapped to human chromosome 2p25.3. The encoded protein is a member of the human peroxidases family. TPO is a ~100 kDa protein with 933-amino acids. Mature TPO is characterized with a short transmembrane and cytoplasmic regions, as well as a dominant ectodomain, containing MPO-like, complement control protein (CCP)-like and epidermal growth factor (EGF)-like domains.

Immunogen

Peptide with sequence C-TRHVIQVSNEVVTDD, from the internal region of the protein sequence according toNP_000538.3; NP_783650.1; NP_783652.1; NP_783653.1

Biochem/physiol Actions

Thyroid peroxidase (TPO) is involved in the biosynthesis of thyroid hormones T3 and T4. It catalyzes the oxidation of the inorganic iodide and coupling of iodinated tyrosine to produce thyroxine and triiodothyronine. Mutation in the gene increases the risk of susceptibility to autoimmune thyroiditis in patients with allergic rhinitis and asthma. Genetic deficiency of TPO is associated with the development of congenital hypothyroidism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum Anti-TPO and TPO Gene Polymorphism as a Predictive Factor for Hidden Autoimmune Thyroiditis in Patient with Bronchial Asthma and Allergic Rhinitis.
El RS, et al.
The Egyptian Journal of Immunology / Egyptian Association of Immunologists, 23(1), 77-86 (2016)
Thyroid peroxidase (TPO) expressed in thyroid and breast tissues shows similar antigenic properties.
Godlewska M, et al.
PLoS ONE, 12(6) (2017)
Identification and characterization of VPO1, a new animal heme-containing Peroxidase
Cheng G, et al.
Free Radical Biology & Medicine, 45(12), 1682-1694 (2008)
Triallelic patterns in STR loci used for paternity analysis: Evidence for a duplication in chromosome 2 containing the TPOX STR locus
Lukka M, et al.
Forensic Science International, 164(1), 3-9 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service