Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501370

Sigma-Aldrich

Anti-SNAI1 (N-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SLUGH2, Anti-SNA, Anti-SNAH, Anti-dJ710H13.1, Snail homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€477.00

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

indirect ELISA: suitable

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAI1(6615)

General description

Snail family transcriptional repressor 1 (SNAIL) is a transcription factor and the gene encoding it is localized on human chromosome 20q13.13.

Immunogen

Peptide with sequence RKPSDPNRKPNY-C, from the N-terminal region of the protein sequence according to NP_005976.2

Biochem/physiol Actions

Snail family transcriptional repressor 1 (SNAIL) acts as a repressor of genes associated with the epithelial phenotype like E-cadherin and increases the expression of genes linked with the mesenchymal phenotype. It has a role in epithelial-mesenchymal transition. The protein aids in G1 transition (early to late) in the cell cycle. SNAIL also modulates cell-matrix adhesion.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TP53 Mutation, Epithelial-Mesenchymal Transition, and StemlikeFeatures in Breast Cancer Subtypes
Danila Coradini
Journal of Biomedicine and Biotechnology (2012)
CRISPR/Cas9n-Mediated Deletion of the Snail 1Gene (SNAI1) Reveals Its Role in Regulating Cell Morphology, Cell-Cell Interactions, and Gene Expression in Ovarian Cancer (RMG-1) Cells.
Haraguchi M
PLoS ONE (2015)
SNAIL transcription factor increases the motility and invasive capacity of prostate cancer cells.
Osorio LA
Molecular Medicine Reports (2016)
Jessica Catapano et al.
Cellular & molecular biology letters, 27(1), 100-100 (2022-11-20)
Metformin is an inhibitor of oxidative phosphorylation that displays an array of anticancer activities. The interference of metformin with the activity of multi-drug resistance systems in cancer cells has been reported. However, the consequences of the acquired chemoresistance for the
Paweł Kochanowski et al.
International journal of molecular sciences, 22(8) (2021-05-01)
Glioblastoma multiforme (GBM) recurrences after temozolomide (TMZ) treatment result from the expansion of drug-resistant and potentially invasive GBM cells. This process is facilitated by O6-Methylguanine-DNA Methyltransferase (MGMT), which counteracts alkylating TMZ activity. We traced the expansion of invasive cell lineages

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service