Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501246

Sigma-Aldrich

Anti-Connexin 43 Antibody

goat polyclonal

Synonym(s):

Anti-CX43, Anti-DFNB38, Anti-GJAL, Anti-ODDD, gap junction protein, alpha 1, 43kDa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-GJA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJA1(2697)

Immunogen

Peptide with sequence CQPFDFPDDNQNSKK, from the internal region of the protein sequence according to NP_000156.1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenghai Zhou et al.
Experimental and therapeutic medicine, 11(6), 2139-2146 (2016-06-11)
The aim of the present study was to investigate the association between connexin (Cx)43 levels and alterations in gap junctional mediation of intercellular communication in overactive bladder syndrome (OAB), and to examine the effects of connexin inhibitor on this condition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service