Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501042

Sigma-Aldrich

Anti-TIP30/CC3 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HIV-1 Tat interactive protein 2, Anti-HTATIP2, Anti-Tat-interacting protein (30 kDa)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€421.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTATIP2(10553)

General description

Tat-interacting protein 30 (TIP30), also called CC3/ HIV-1 tat interactive protein 2 (HTATIP2), is a putative tumor suppressor. It is encoded by the gene mapped to human chromosome 11p15.1. TIP30 is primarily localized in cytoplasm and it belongs to short-chain dehydrogenases/reductase (SDR) family. The encoded protein is extensively expressed in various human tissues, such as the heart, brain, lung, kidney and pancreas.

Immunogen

Peptide with sequence AETEALSKLREDFR-C from the N Terminus of the protein sequence according to NP_006401.

Biochem/physiol Actions

Tat-interacting protein 30 (TIP30) has an ability to promote apoptosis and suppress metastasis and angiogenesis. It is also implicated in regulation of DNA repair and tumor cell metabolism. Down-regulated expression of the gene is associated with the development of various cancers, including melanoma, breast cancer, neuroblastoma, glioblastoma, colon cancer, lung cancer and hepatocellular carcinoma. Elevated expression of the gene in glioma cells inhibits tumor proliferation, metastasis and migration. Thus, TIP30 can be considered as a potent target for treating glioma patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TIP30 Deficiency Increases Susceptibility to Tumorigenesis
Ito M, et al.
Cancer Research, 63(24), 8763-8767 (2003)
Overexpression of TIP30 inhibits the growth and invasion of glioma cells
Hu Y, et al.
Molecular Medicine Reports, 13(1), 605-612 (2016)
TIP30: a novel tumor-suppressor gene.
Yu X, et al.
Oncology Research, 22(5-6), 339-348 (2015)
Tat-Interacting Protein 30 (TIP30) Expression Serves as a New Biomarker for Tumor Prognosis: A Systematic Review and Meta-Analysis
Xu T, et al.
PLoS ONE, 11(12), e0168408-e0168408 (2016)
Xin Tong et al.
The American journal of pathology, 174(5), 1931-1939 (2009-04-08)
The HIV Tat-interacting protein (TIP30), also called CC3 or HTIP2, is encoded by Tip30, a putative tumor-suppressor gene located on human chromosome 11p15.1. In this study, we investigated the role of TIP30 in the progression and metastasis of lung cancer.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service