Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500715

Sigma-Aldrich

Anti-OGT antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FLJ23071, Anti-HRNT1, Anti-MGC22921, Anti-O-GLCNAC, Anti-O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1323


Select a Size

Change View
100 μG
€421.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€421.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1323

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OGT(8473)

General description

O-linked N-acetylglucosamine (GlcNAc) transferase (OGT) has 13.5 tetratricopeptide-repeats (TPR) and a globular catalytic domain. The gene encoding this protein is localized on human chromosome Xq13.1.

Immunogen

Peptide with sequence C-YEHPKDLKLSDGR from the internal region of the protein sequence according to NP_858058.1; NP_858059.1.

Biochem/physiol Actions

O-linked N-acetylglucosamine (GlcNAc) transferase (OGT) has a role in the addition of O-linked N-acetylglucosamine (O-GlcNAc) to serine/threonine residues in the protein of interest. It is crucial for O-GlcNAcylation of RNA polymerase II. Absence of OGT has been shown to reduce transcription and pol II promoter occupancy. The gene is upregulated in breast cancer tissues, colon cancer, lung cancer and laryngeal cancer. This protein might be associated with tumorigenesis and metastasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silencing ?-linked N-acetylglucosamine transferase induces apoptosis in human gastric cancer cells through PUMA and caspase-3 pathways.
Wen T
Oncology Reports (2015)
Human RNA Polymerase II Promoter Recruitment in Vitro Is Regulated by O-Linked N-Acetylglucosaminyltransferase (OGT).
Lewis BA
The Journal of Biological Chemistry (2016)
Distinct OGT-Binding Sites Promote HCF-1 Cleavage.
Bhuiyan T
PLoS ONE (2015)
Human O-GlcNAc transferase (OGT): genomic structure, analysis of splice variants, fine mapping in Xq13.1.
Nolte D and Muller U
Mammalian Genome (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service