Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500703

Sigma-Aldrich

Anti-NOVA1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neuro-oncological ventral antigen 1, Anti-Paraneoplastic Ri antigen, Anti-Ventral neuron-specific protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOVA1(4857)

General description

NOVA1 (Neuro-oncological ventral antigen 1) is a neuron-specific nuclear RNA-binding protein expressed in brain, specifically in the hippocampus, hypothalamus, cingulate cortex, and medial habenular nucleus.

Immunogen

Peptide with sequence C-REMPQNVAKTEPVS from the internal region of the protein sequence according to NP_002506.2 ; NP_006480.2 ; NP_006482.1.

Application

Anti-NOVA1 antibody produced in goat is suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) and western blot analysis.

Biochem/physiol Actions

NOVA1 (Neuro-oncological ventral antigen 1) is associated with several post-transcriptional regulation of RNA metabolism including RNA splicing, editing to transport, localization, and degradation. It may be involved in mediating neuronal responsiveness. It encodes a protein, which has homology with the RNA-binding protein hnRNP K, the yeast splicing protein MER1.Since, it has some homology with hnRNP K, it may influence the regulation of RNA splicing or metabolism in developing neurons. The importance of NOVA1 as prognostic marker has been reported in HCC (Hepatocellular carcinoma). Alteration in gene causes a disorder associated with breast cancer and motor dysfunction i.e. paraneoplastic opsoclonus myoclonus ataxia (POMA).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Buckanovich et al.
Molecular and cellular biology, 17(6), 3194-3201 (1997-06-01)
Nova-1, an autoantigen in paraneoplastic opsoclonus myoclonus ataxia (POMA), a disorder associated with breast cancer and motor dysfunction, is a neuron-specific nuclear RNA binding protein. We have identified in vivo Nova-1 RNA ligands by combining affinity-elution-based RNA selection with protein-RNA
Yi-An Zhang et al.
PloS one, 9(3), e90955-e90955 (2014-03-13)
Neuro-oncological ventral antigen 1 (Nova1) is a neuron-specific RNA-binding protein in human paraneoplastic opsoclonus-myoclonus ataxia accompanying with malignant tumors, but its role in hepatocellular carcinoma (HCC) remains elusive. In this study, we found that overexpressed intratumoral Nova1 was associated with
R J Buckanovich et al.
Neuron, 11(4), 657-672 (1993-10-01)
Paraneoplastic opsoclonus-ataxia, a disorder of motor control, develops in breast or lung cancer patients who harbor an antibody (Ri) that recognizes their tumors and a nuclear neuronal protein of 55 kd. We have characterized a gene, Nova, encoding an antigen
Hualing Li et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 61(1), 45-54 (2012-10-09)
The present study aimed to evaluate the expression of neuro-oncological ventral antigen 1 (Nova1) in cerebral ischemia/reperfusion (I/R) insults by immunohistochemistry. The focal cerebral I/R model was induced by right middle cerebral artery occlusion (MCAO) for 120 min followed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service