Skip to Content
Merck
All Photos(6)

Key Documents

SAB2500451

Sigma-Aldrich

Anti-GAPDH Antibody

goat polyclonal

Synonym(s):

Anti-G3PD, Anti-GAPD, Anti-Glyceraldehyde-3-phosphate dehydrogenase, Anti-HGNC:4141, Anti-MGC88685

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IF
IHC
WB
clone:
polyclonal
species reactivity:
human
citations:
11
technique(s):
ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

Product Name

Anti-GAPDH (Internal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

General description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) localizes in the cytoplasm but can be translocated to the nucleus depending on cellular conditions. It is a tetramer containing identical chains. The gene encoding this protein is localized on human chromosome 12p13.

Immunogen

Peptide with sequence C-GVNHEKYDNSLK from the internal region of the protein sequence according to NP_002037.2.

Application

Anti-GAPDH (Internal) antibody produced in goat has been used in Western blotting.

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) catalyzes the reversible oxidative phosphorylation of glyceraldehyde-phosphate, which is a critical energy-yielding step in carbohydrate metabolism. It binds to several proteins including actin, tubulin, amyloid precursor, polyglutamine peptides, DRPLA (dentatorubral-pallidoluysian atrophy) and huntingtin. Phosphorylated GAPDH associates with cytoskeletal elements and controls microtubule dynamics in the early secretory pathway. GAPDH is also a component of the functional GAIT (interferon-γ-activated inhibitor of translation) mRNP (messenger ribonucleoprotein). GAPDH expression is dysregulated during melanoma progression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WT1 shRNA delivery using transferrin-conjugated PEG liposomes in an in vivo model of melanoma
Experimental and Therapeutic Medicine (2014)
Mutant huntingtin: nuclear translocation and cytotoxicity mediated by GAPDH.
Bae BL
Proceedings of the National Academy of Sciences of the USA (2006)
Noncanonical function of glutamyl-prolyl-tRNA synthetase: gene-specific silencing of translation.
Sampath P
Cell (2004)
Deregulation of glyceraldehyde-3-phosphate dehydrogenase expression during tumor progression of human cutaneous melanoma.
Ramos D
Anticancer Research (2015)
AIM2 Inflammasome Is Critical for Influenza-Induced Lung Injury and Mortality
Hongbo Zhang
Journal of Immunology (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service