Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500188

Sigma-Aldrich

Anti-Calretinin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ANX2, Anti-ANXA2, Anti-Annexin A2, Anti-LIP2, Anti-LPC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€477.00

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZMS1073


Select a Size

Change View
100 μG
€477.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€477.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZMS1073

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, bovine, mouse, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA2(302)

Immunogen

Peptide with sequence STVHEILCKLSLEGD from the N Terminus of the protein sequence according to NP_001002858.1; NP_004030.1; NP_001002857.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher M Whitaker et al.
Frontiers in cellular neuroscience, 15, 662329-662329 (2021-05-25)
Rod and cone pathways are segregated in the first stage of the retina: cones synapse with both ON- and OFF-cone bipolar cells while rods contact only rod bipolar cells. However, there is an exception to this specific wiring in that
Luis Pérez de Sevilla Müller et al.
The Journal of comparative neurology, 527(1), 133-158 (2017-05-05)
Amacrine cells are a heterogeneous group of interneurons that form microcircuits with bipolar, amacrine and ganglion cells to process visual information in the inner retina. This study has characterized the morphology, neurochemistry and major cell types of a VIP-ires-Cre amacrine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service