Skip to Content
Merck
All Photos(11)

Key Documents

SAB2500041

Sigma-Aldrich

Anti-AIF1/IBA1 (Isoforms 1 and 3) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AIF-1, Anti-Allograft inflammatory factor 1, Anti-IBA1, Anti-IRT-1, Anti-Interferon γ responsive transcript

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€561.00

€561.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1024


Select a Size

Change View
100 μG
€561.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€561.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1024

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AIF1(199)

Immunogen

Peptide with sequence C-TGPPAKKAISELP from the C Terminus of the protein sequence according to NP_116573.1; NP_001614.3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paramita Basu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(47), 9827-9843 (2021-09-18)
Latent sensitization (LS) of pain engages pronociceptive signaling pathways in the dorsal horn that include NMDA receptor (NMDAR)→adenylyl cyclase-1 (AC1)→protein kinase A (PKA), and exchange proteins directly activated by cyclic AMP (Epacs). To determine whether these pathways operate similarly between
Martin E Gosnell et al.
Neurobiology of disease, 160, 105528-105528 (2021-10-10)
Our understanding of chronic pain and the underlying molecular mechanisms remains limited due to a lack of tools to identify the complex phenomena responsible for exaggerated pain behaviours. Furthermore, currently there is no objective measure of pain with current assessment
Blanca Lizarbe et al.
Frontiers in neuroscience, 12, 985-985 (2019-01-24)
Metabolic syndrome and diabetes impact brain function and metabolism. While it is well established that rodents exposed to diets rich in saturated fat develop brain dysfunction, contrasting results abound in the literature, likely as result of exposure to different high-fat
Daniela F Santos et al.
International journal of molecular sciences, 22(21) (2021-11-14)
(Poly)phenol-derived metabolites are small molecules resulting from (poly)phenol metabolization after ingestion that can be found in circulation. In the last decade, studies on the impact of (poly)phenol properties in health and cellular metabolism accumulated evidence that (poly)phenols are beneficial against
Temporal tracking of microglial and monocyte single-cell transcriptomics in lethal flavivirus infection.
Spiteri, et al.
Acta Neuropathologica Communications, 11, 60-60 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service