Skip to Content
Merck
All Photos(2)

Documents

SAB2108578

Sigma-Aldrich

Anti-SLC22A3

affinity isolated antibody

Synonym(s):

Anti- EMTH, Anti- OCT3, Anti-EMT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_021977

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC22A3(6581)

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC22A3

Biochem/physiol Actions

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. SLC22A3 contains twelve putative transmembrane domains and is a plasma integral membrane protein.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein.

Sequence

Synthetic peptide located within the following region: PDHYWCRGPSAAALAERCGWSPEEEWNRTAPASRGPEPPERRGRCQRYLL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metee Jinakote et al.
Fundamental & clinical pharmacology, 37(4), 833-842 (2023-02-28)
Buspirone, a cationic drug, is an anxiolytic and antidepressant drug. However, whether buspirone and its metabolites are interacted with organic cationic transporter remains uncertain. In this study, we examined the interaction of buspirone and its major metabolites 1-(2-pyrimidinyl)piperazine (1-PP) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service