Skip to Content
Merck
All Photos(1)

Key Documents

SAB2108568

Sigma-Aldrich

Anti-FAM107A

IgG fraction of antiserum

Synonym(s):

Anti- FLJ30158, Anti- FLJ45473, Anti- TU3A, Anti-DRR1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

guinea pig, bovine, human, mouse, dog, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_007177

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM107A(11170)

General description

FAM107A (family with sequence similarity 107 member A) is expressed at high level in the brain and heart. It has a nuclear localization signal (NLS) and a coiled domain. FAM107A has 144 amino acids and is located on human chromosome 3p14.3.

Immunogen

Synthetic peptide directed towards the middle region of human FAM107A

Application

Anti-FAM107A has been used in the quantification of DRR1 protein levels.[1]

Biochem/physiol Actions

FAM107A (family with sequence similarity 107 member A) participates in neuronal cell survival. Overexpression of FAM107A has the ability to repress the development of tumor cells. It plays a major role in the progression of the embryo. FAM107A participates in cell invasion. This protein modulates FA dynamics and cell movement.
When FAM107A is transfected into cell lines in which it is not expressed, it suppresses cell growth. It may play a role in tumor development.

Sequence

Synthetic peptide located within the following region: RLQCPFEQELLRRQQRLNQLEKPPEKEEDHAPEFIKVRENLRRIATLTSE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FAM107A (family with sequence similarity 107, member A)
Kadomatsu K and Mu P
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2011)
Tanja Jene et al.
Psychoneuroendocrinology, 91, 149-158 (2018-03-21)
Understanding the neurobiological mechanisms underlying the response to an acute stressor may provide novel insights into successful stress-coping strategies. Acute behavioral stress-effects may be restricted to a specific time window early after stress-induction. However, existing behavioral test batteries typically span

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service