Skip to Content
Merck
All Photos(3)

Key Documents

SAB2108414

Sigma-Aldrich

Anti-B3GALT2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€489.00

€489.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

49kDa

species reactivity

horse, mouse, rat, dog, guinea pig, human, goat, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B3GALT2(8707)
mouse ... B3galt2(26878)

Immunogen

Synthetic peptide directed towards the C terminal of human B3GALT2

Biochem/physiol Actions

B3galt2 is beta-1,3-galactosyltransferase that transfers galactose from UDP-galactose to substrates with a terminal beta-N-acetylglucosamine (beta-GlcNAc) residue. B3galt2 can also utilize substrates with a terminal galactose residue, albeit with lower efficiency. B3galt2 is involved in the biosynthesis of the carbohydrate moieties of glycolipids and glycoproteins.

Sequence

Synthetic peptide located within the following region: RVDPVPPPNEFVFNHWRVSYSSCKYSHLITSHQFQPSELIKYWNHLQQNK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xun Guo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 36(10), e22542-e22542 (2022-09-13)
Ischemic stroke is one of the major causes of morbidity and mortality. The β-1, 3-galactosyltransferase 2 (B3galt2), a member of β-1, 3-galactosyltransferase family, is playing a vital role in the pathological process of cerebral ischemic injury, but its underlying mechanisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service