Skip to Content
Merck
All Photos(3)

Key Documents

SAB2108306

Sigma-Aldrich

Anti-SOX4 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47kDa

species reactivity

mouse, guinea pig, rabbit, dog, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX4(6659)

Immunogen

Synthetic peptide directed towards the N terminal region of human SOX4

Biochem/physiol Actions

SOX4 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. It may act as a transcriptional regulator after forming a protein complex with other proteins, such as syndecan binding protein (syntenin). The protein may function in the apoptosis pathway leading to cell death as well as to tumorigenesis and may mediate downstream effects of parathyroid hormone (PTH) and PTH-related protein (PTHrP) in bone development. The solution structure has been resolved for the HMG-box of a similar mouse protein. This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins, such as syndecan binding protein (syntenin). The protein may function in the apoptosis pathway leading to cell death as well as to tumorigenesis and may mediate downstream effects of parathyroid hormone (PTH) and PTH-related protein (PTHrP) in bone development. The solution structure has been resolved for the HMG-box of a similar mouse protein. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: MVQQTNNAENTEALLAGESSDSGAGLELGIASSPTPGSTASTGGKADDPS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Bai et al.
Experimental and therapeutic medicine, 16(3), 2235-2242 (2018-09-07)
Accumulating evidence has demonstrated that long noncoding RNAs (lncRNAs) serve important roles in tumor development and progression. However, whether lncRNA EZR-AS1 is associated with breast cancer (BC) progression remains unclear. In the present study, reverse transcription-quantitative polymerase chain reaction analysis
Bingting Pan et al.
International journal of molecular medicine, 40(6), 1691-1698 (2017-10-19)
Bronchopulmonary dysplasia (BPD) is currently the most common severe complication in premature infants and is characterized by the arrest of alveolar and vascular growth. Alveolar type II cells play an important role in the pathological foundation of BPD. An association of
Xueran Chen et al.
Neuro-oncology, 25(1), 82-96 (2022-06-22)
Glioblastoma multiforme and other solid malignancies are heterogeneous, containing subpopulations of tumor cells that exhibit stem characteristics. Oct4, also known as POU5F1, is a key transcription factor in the self-renewal, proliferation, and differentiation of stem cells. Although it has been
Peng Zhang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 42(1), 242-253 (2017-05-24)
Recently, microRNAs (miRNA) have been identified as novel regulators in Chondrosarcoma (CHS). This study was aimed to identify the roles of miR-129-5p-5p in regulation of SOX4 and Wnt/β-catenin signaling pathway, as well as cell proliferation and apoptosis in chondrosarcomas. Tissue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service