Skip to Content
Merck
All Photos(7)

Key Documents

SAB2108147

Sigma-Aldrich

Anti-GSR antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€454.00

€454.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€454.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€454.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56kDa

species reactivity

mouse, human, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSR(2936)

Immunogen

Synthetic peptide directed towards the N terminal region of human GSR

Biochem/physiol Actions

GSR belongs to the class-I pyridine nucleotide-disulfide oxidoreductase family. It maintains high levels of reduced glutathione in the cytosol. Both glutathione and glutathione reductase are inducible by D3T, and that upregulation of GSH biosynthesis underlies D3T-mediated cytoprotection against ROS/RNS-elicited injury to human vascular smooth muscle cells.

Sequence

Synthetic peptide located within the following region: PTIEVSGKKYTAPHILIATGGMPSTPHESQIPGASLGITSDGFFQLEELP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heloina Nathalliê Mariano da Silva et al.
Frontiers in physiology, 12, 649793-649793 (2021-05-14)
The mdx mouse phenotype aggravated by chronic exercise on a treadmill makes this murine model more reliable for the study of muscular dystrophy. Thus, to better assess the Tempol effect on dystrophic pathways, the analyses in this study were performed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service