Skip to Content
Merck
All Photos(1)

Documents

SAB2105049

Sigma-Aldrich

Anti-RAGE, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MOK, Anti-RAGE1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

dog, human, bovine, rat, mouse, horse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAGE(5891)

Immunogen

Synthetic peptide directed towards the N terminal region of human RAGE

Sequence

Synthetic peptide located within the following region: MKQRFESIEQVNNLREIQALRRLNPHPNILMLHEVVFDRKSGSLALICEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Wang et al.
PloS one, 12(7), e0181248-e0181248 (2017-07-15)
The NLRP3 inflammasome plays an important role in the pathogenesis of inflammation in diabetic nephropathy (DN). Pioglitazone (PIO) has been found to exert an anti-inflammatory effect in patients with diabetes mellitus, but it is still unclear whether PIO exhibits a
Beibing Di et al.
Peptides, 112, 125-132 (2018-12-05)
Vascular smooth muscle cell (VSMC) phenotype transition is involved in diabetes-associated cardiovascular diseases. The mechanism of VSMCs phenotypic transition in T2DM was still unclear. Rat coronary artery SMCs were pretreated with liraglutide alone, liraglutide and H89(a PKA inhibitor), neutralizing anti-RAGE
Yong-Gang Fan et al.
EBioMedicine, 45, 393-407 (2019-07-16)
Recent studies have revealed that vitamin D deficiency may increase the risk of Alzheimer's disease, and vitamin D supplementation may be effective strategy to ameliorate the neurodegenerative process in Alzheimer's disease patients. Paricalcitol (PAL), a low-calcemic vitamin D receptor agonist
Yong-Gang Fan et al.
British journal of pharmacology, 181(6), 896-913 (2023-06-13)
Overexpression of astrocytic lactoferrin (Lf) was observed in the brain of Alzheimer's disease (AD) patients, whereas the role of astrocytic Lf in AD progression remains unexplored. In this study, we aimed to evaluate the effects of astrocytic Lf on AD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service