Skip to Content
Merck
All Photos(2)

Documents

SAB2102222

Sigma-Aldrich

Anti-SLC6A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GABATHG, Anti-GABATR, Anti-GAT1, Anti-Solute carrier family 6 (neurotransmitter transporter, GABA), member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67 kDa

species reactivity

mouse, guinea pig, rat, dog, bovine, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC6A1(6529)

Immunogen

Synthetic peptide directed towards the middle region of human SLC6A1

Biochem/physiol Actions

SLC6A1 terminates the action of GABA by its high affinity sodium-dependent reuptake into presynaptic terminals. This protein is the target of psychomotor stimulants such as amphetamines or cocaine.

Sequence

Synthetic peptide located within the following region: CCINSCTSMFAGFVIFSIVGFMAHVTKRSIADVAASGPGLAFLAYPEAVT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Kim et al.
Neuroscience, 262, 129-142 (2014-01-15)
GABA is a predominant inhibitory neurotransmitter in the CNS. Released GABA is removed from the synaptic cleft by two GABA transporters (GATs), GAT-1 and GAT-3, and their dysfunction affects brain functions. The present study aimed to reveal the ontogeny of

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service