Skip to Content
Merck
All Photos(2)

Documents

SAB2101082

Sigma-Aldrich

Anti-HS6ST3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp761K2315, Anti-Heparan sulfate 6-O-sulfotransferase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

rat, dog, guinea pig, rabbit, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HS6ST3(266722)

Immunogen

Synthetic peptide directed towards the C terminal region of human HS6ST3

Biochem/physiol Actions

Heparan sulfate (HS) sulfotransferases, such as HS6ST3, modify HS to generate structures required for interactions between HS and a variety of proteins. These interactions are implicated in proliferation and differentiation, adhesion, migration, inflammation, blood coagulation, and other diverse processes.Heparan sulfate (HS) sulfotransferases, such as HS6ST3, modify HS to generate structures required for interactions between HS and a variety of proteins. These interactions are implicated in proliferation and differentiation, adhesion, migration, inflammation, blood coagulation, and other diverse processes (Habuchi et al., 2000 [PubMed 10644753]).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: TKQLEHQRDRQKRREERRLQREHRDHQWPKEDGAAEGTVTEDYNSQVVRW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Li et al.
Nature communications, 9(1), 2829-2829 (2018-07-22)
Recent studies suggest the emerging roles of armadillo (ARM) family proteins in tumor progression. However, the functions and underlying mechanisms of ARM members in tumorigenesis and aggressiveness of neuroblastoma (NB) remain to be determined. Herein, we identify armadillo repeat containing 12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service