Skip to Content
Merck
All Photos(4)

Key Documents

SAB2100593

Sigma-Aldrich

Anti-DLL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DELTA1, Anti-Delta, Anti-Delta-like 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

78 kDa

species reactivity

rat, pig, bovine, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLL1(28514)

Immunogen

Synthetic peptide directed towards the N terminal region of human DLL1

Biochem/physiol Actions

DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication.DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: GSRCALALAVLSALLCQVWSSGVFELKLQEFVNKKGLLGNRNCCRGGAGP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylwia Lustofin et al.
International journal of molecular sciences, 23(4) (2022-02-27)
Delta/Serrate/LAG-2 (DSL) proteins, which serve as ligands for Notch receptors, mediate direct cell-cell interactions involved in the determination of cell fate and functioning. The present study aimed to explore the role of androgens and estrogens, and their receptors in the
Alicja Kamińska et al.
Reproductive biology and endocrinology : RB&E, 18(1), 30-30 (2020-04-18)
Onset of spermatogenesis at puberty is critically dependent on the activity of hypothalamic-pituitary-gonadal axis and testosterone production by Leydig cells. The aim of this study was to examine whether activation of Notch receptors and expression of Notch ligands and effector

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service