Skip to Content
Merck
All Photos(4)

Key Documents

SAB2100040

Sigma-Aldrich

Anti-ACTN3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Actinin, α3, Anti-MGC117002, Anti-MGC117005

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

103 kDa

species reactivity

dog, rat, guinea pig, mouse, bovine, rabbit, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTN3(89)

Immunogen

Synthetic peptide directed towards the N terminal region of human ACTN3

Biochem/physiol Actions

Alpha-actinin is an actin-binding protein with multiple roles in different cell types. This protein expression is limited to skeletal muscle. It is localized to the Z-disc and analogous dense bodies, where it helps to anchor the myofibrillar actin filaments.Alpha-actinin is an actin-binding protein with multiple roles in different cell types. This gene expression is limited to skeletal muscle. It is localized to the Z-disc and analogous dense bodies, where it helps to anchor the myofibrillar actin filaments. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: VQNFHTSWKDGLALCALIHRHRPDLIDYAKLRKDDPIGNLNTAFEVAEKY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Gagaoua et al.
Food chemistry, 250, 245-252 (2018-02-08)
The validation of biomarkers and tools for the prediction of beef texture remains a challenging task. In this study, reverse phase protein arrays (RPPA) quantified 29 protein biomarkers in the m. Longissimus thoracis of Charolais cattle sampled early post-mortem. Myosin
Brigitte Picard et al.
Journal of proteomics, 273, 104792-104792 (2022-12-20)
We aimed to evaluate the relationships between meat or carcass properties and the abundance of 29 proteins quantified in two muscles, Longissimus thoracis and Rectus abdominis, of Rouge des Prés cows. The relative abundance of the proteins was evaluated using
Mohammed Gagaoua et al.
Meat science, 145, 308-319 (2018-07-18)
Reverse Phase Protein Arrays (RPPA) were applied for the quantification and validation of protein biomarkers of beef qualities on M. longissimus thoracis sampled early post-mortem from young Charolais bulls. pHu was related to six proteins, three of which are glycolytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service