Skip to Content
Merck
All Photos(1)

Key Documents

SAB1412433

Sigma-Aldrich

ANTI-MUC2 antibody produced in mouse

clone 1H2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MLP, MUC2, SMUC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€494.00

€494.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€494.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€494.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H2, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC2(4583)

General description

This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. (provided by RefSeq)

Immunogen

MUC2 (NP_002448, 5081 a.a. ~ 5179 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TTEVSYAGCTKTVLMNHCSGSCGTFVMYSAKAQALDHSCSCCKEEKTSQREVVLSCPNGGSLTHTYTHIESCQCQDTVCGLPTGTSRRARRSPRHLGSG*

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service