Skip to Content
Merck
All Photos(3)

Key Documents

SAB1412387

Sigma-Aldrich

ANTI-ARHGEF1 antibody produced in mouse

clone 2D2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ARHGEF1, GEF1, LBCL2, LSC, P115-RHOGEF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€494.00

€494.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€494.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€494.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D2, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.41 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF1(9138)

General description

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. (provided by RefSeq)

Immunogen

ARHGEF1 (AAH34013.2, 830 a.a. ~ 927 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service