Skip to Content
Merck
All Photos(2)

Documents

SAB1411908

Sigma-Aldrich

ANTI-MAGEA9 antibody produced in mouse

clone 2C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MAGE9, MAGEA9, MGC8421

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C2, monoclonal

form

buffered aqueous solution

mol wt

antigen 60.39 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGEA9(4108)

Related Categories

General description

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. (provided by RefSeq)

Immunogen

MAGEA9 (AAH02351.1, 1 a.a. ~ 315 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSLEQRSPHCKPDEDLEAQGEDLGLMGAQEPTGEEEETTSSSDSKEEEVSAAGSSSPPQSPQGGASSSISVYYTLWSQFDEGSSSQEEEEPSSSVDPAQLEFMFQEALELKVAELVHFLLHKYRVKEPVTKAEMLESVIKNYKRYFPVIFGKASEFMQVIFGTDVKEVDPAGHSYILVTALGLSCDSMLGDGHSMPKAALLIIVLGVILTKDNCAPEEVIWEALSVMGVYVGKEHMFYGEPRKLLTQDWVQENYLEYRQVPGSDPAHYEFLWGSKAHAETSYEKVINYLVMLNAREPICYPSLYEEVLGEEQEGV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service