Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411324

Sigma-Aldrich

Anti-TAF10 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

TAF2A, TAF2H, TAFII30

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAF10(6881)

General description

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs. (provided by RefSeq)

Immunogen

TAF10 (NP_006275.1, 1 a.a. ~ 218 a.a) full-length human protein.

Sequence
MSCSGSGADPEAAPASAASAPGPAPPVSAPAALPSSTAAENKASPAGTAGGPGAGAAAGGTGPLAARAGEPAERRGAAPVSAGGAAPPEGAISNGVYVLPSAANGDVKPVVSSTPLVDFLMQLEDYTPTIPDAVTGYYLNRAGFEASDPRIIRLISLAAQKFISDIANDALQHCKMKGTASGSSRSKSKDRKYTLTMEDLTPALSEYGINVKKPHYFT

Biochem/physiol Actions

TAF10 plays a major role in transcription by binding to the TBP-associated factors (TAFs). It interacts with the AF-2-containing region E of the human estrogen receptor (ER). In conjugation with high-mobility group (HMG) protein HMG-1, TAF10 facilitates estrogen receptor-mediated transcriptional activation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Van Der Knaap et al.
The Biochemical journal, 345 Pt 3, 521-527 (2000-01-22)
The TATA-binding protein (TBP) plays a central role in eukaryotic transcription and forms protein complexes with TBP-associated factors (TAFs). The genes encoding TAF(II) proteins frequently map to chromosomal regions altered in human neoplasias. TAF(II)170 of B-TFIID is a member of
C S Verrier et al.
Molecular endocrinology (Baltimore, Md.), 11(8), 1009-1019 (1997-07-01)
The estrogen receptor (ER) belongs to a family of ligand-inducible nuclear receptors that exert their effects by binding to cis-acting DNA elements in the regulatory region of target genes. The detailed mechanisms by which ER interacts with the estrogen response
X Jacq et al.
Cell, 79(1), 107-117 (1994-10-07)
We showed previously that coactivators mediating stimulation by different activators were associated with the TATA-binding protein (TBP) in distinct TFIID complexes. We have characterized a human TBP-associated factor (TAF), hTAFII30, associated with a subset of TFIID complexes. hTAFII30 interacts with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service