Skip to Content
Merck
All Photos(3)

Key Documents

SAB1411320

Sigma-Aldrich

Anti-SUOX antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53.9 kDa

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUOX(6821)

General description

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins. (provided by RefSeq)

Immunogen

SUOX (ENSP00000348440, 1 a.a. ~ 488 a.a) full-length human protein.

Sequence
MGTLLGLGAVLAYQDHRCRAAQESTHIYTKEEVSSHTSPETGIWVTLGSEVFDVTEFVDLHPGGPSKLMLAAGGPLEPFWALYAVHNQSHVRELLAQYKIGELNPEDKVAPTVETSDPYADDPVRHPALKVNSQRPFNAEPPPELLTENYITPNPIFFTRNHLPVPNLDPDTYRLHVVGAPGGQSLSLSLDDLHNFPRYEITVTLQCAGNRRSEMTQVKEVKGLEWRTGAISTARWAGARLCDVLAQAGHQLCETEAHVCFEGLDSDPTGTAYGASIPLARAMDPEAEVLLAYEMNGQPLPRDHGFPVRVVVPGVVGARHVKWLGRVSVQPEESYSHWQRRDYKGFSPSVDWETVDFDSAPSIQELPVQSAITEPRDGETVESGEVTIKGYAWSGGGRAVIRVDVSLDGGLTWQVAKLDGEEQRPRKAWAWRLWQLKAPVPAGQKELNIVCKAVDDGYNVQPDTVAPIWNLRGVLSNAWHRVHVYVSP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service