Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411280

Sigma-Aldrich

Anti-PINK1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BRPK, FLJ27236, PARK6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)

General description

PTEN induced putative kinase 1 (PINK1) is a serine/threonine mitochondrial kinase. The 581 amino acid protein has an amino terminal mitochondrial target sequence, a putative transmembrane domain, a kinase domain and an autophosphorylation-regulating carboxy terminal domain. The gene encoding it is localized on human chromosome 1p36.12.
This gene encodes a serine/threonine protein kinase that localizes to mitochondria. It is thought to protect cells from stress-induced mitochondrial dysfunction. Mutations in this gene cause one form of autosomal recessive early-onset Parkinson disease. (provided by RefSeq)

Immunogen

PINK1 (AAH28215.1, 1 a.a. ~ 581 a.a) full-length human protein.

Sequence
MAVRQALGRGLQLGRALLLRFTGKPGRAYGLGRPGPAAGCVRGERPGWAAGPGAEPRRVGLGLPNRLRFFRQSVAGLAARLQRQFVVRAWGCAGPCGRAVFLAFGLGLGLIEEKQAESRRAVSACQEIQAIFTQKSKPGPDPLDTRRLQGFRLEEYLIGQSIGKGCSAAVYEATMPTLPQNLEVTKSTGLLPGRGPGTSAPGEGQERAAGAPAFPLAIKMMWNISAGSSSEAILNTMSQELVPASRVALAGEYGAVTYRKSKRGPKQLAPHPNIIRVLRAFTSSVPLLPGALVDYPDVLPSRLHPEGLGHGRTLFLVMKNYPCTLRQYLCVNTPSPRLAAMMLLQLLEGVDHLVQQGIAHRDLKSDNILVELDPDGCPWLVIADFGCCLADESIGLQLPFSSWYVDRGGNGCLMAPEVSTARPGPRAVIDYSKADAWAVGAIAYEIFGLVNPFYGQGKAHLESRSYQEAQLPALPESVPPDVRQLVRALLQREASKRPSARVAANVLHLSLWGEHILALKNLKLDKMVGWLLQQSAATLLANRLTEKCCVETKMKMLFLANLECETLCQAALLLCSWRAAL

Biochem/physiol Actions

PTEN induced putative kinase 1 (PINK1) has a role in removing damaged mitochondria from cells. Mitochondrial damage triggers the accumulation of the protein in the outer mitochondrial membrane, wherein it undergoes autophosphorylation and dimerizes into a supermolecular protein complex. PINK1 then phosphorylates ubiquitin and tags damaged mitochondria for mitophagy. It negatively modulates glioblastoma growth. Mutations in the PINK1 gene have been associated with recessive, early-onset Parkinson′s disease. The protein is expressed in cancerous cells and has a role in cell survival.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruben K Dagda et al.
The Journal of biological chemistry, 284(20), 13843-13855 (2009-03-13)
Mitochondrial dysregulation is strongly implicated in Parkinson disease. Mutations in PTEN-induced kinase 1 (PINK1) are associated with familial parkinsonism and neuropsychiatric disorders. Although overexpressed PINK1 is neuroprotective, less is known about neuronal responses to loss of PINK1 function. We found
Shinsuke Fujioka et al.
Neuro-degenerative diseases, 10(1-4), 257-260 (2012-01-21)
Major progress in genetic studies of Parkinson's disease (PD) and parkinsonism has been achieved in the last two decades. We provide a brief review of the current status of PARK and non-PARK loci/genes, and discuss two new genes: eIF4G1 and
Rui Zhang et al.
Oncology reports, 37(4), 2137-2146 (2017-03-06)
PTEN-induced putative kinase 1 (PINK1) was identified initially as a gene upregulated in cancer cells which regulates cellular processes of significance in cancer cell biology, including cell survival, stress resistance and the cell cycle. However, the expression and function of
Shiori Akabane et al.
The Journal of biological chemistry, 291(31), 16162-16174 (2016-06-16)
Phosphatase and tensin homolog-induced putative kinase 1 (PINK1), a Ser/Thr kinase, and PARKIN, a ubiquitin ligase, are causal genes for autosomal recessive early-onset parkinsonism. Multiple lines of evidence indicate that PINK1 and PARKIN cooperatively control the quality of the mitochondrial
Sameer Agnihotri et al.
Cancer research, 76(16), 4708-4719 (2016-06-22)
Proliferating cancer cells are characterized by high rates of glycolysis, lactate production, and altered mitochondrial metabolism. This metabolic reprogramming provides important metabolites for proliferation of tumor cells, including glioblastoma. These biological processes, however, generate oxidative stress that must be balanced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service