Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410841

Sigma-Aldrich

Anti-ATP1B3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ATPB-3, CD298, FLJ29027

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 31.5 kDa

species reactivity

human, rat

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP1B3(483)

General description

ATPase Na+/K+ transporting subunit Β 3 (ATP1B3) is a 43kDa protein, encoded by the gene mapped to human chromosome 3q23. The encoded protein is a member of subfamily of Na+/K+ -ATPases.

Immunogen

ATP1B3 (NP_001670.1, 1 a.a. ~ 279 a.a) full-length human protein.

Sequence
MTKNEKKSLNQSLAEWKLFIYNPTTGEFLGRTAKSWGLILLFYLVFYGFLAALFSFTMWVMLQTLNDEVPKYRDQIPSPGLMVFPKPVTALEYTFSRSDPTSYAGYIEDLKKFLKPYTLEEQKNLTVCPDGALFEQKGPVYVACQFPISLLQACSGMNDPDFGYSQGNPCILVKMNRIIGLKPEGVPRIDCVSKNEDIPNVAVYPHNGMIDLKYFPYYGKKLHVGYLQPLVAVQVSFAPNNTGKEVTVECKIDGSANLKSQDDRDKFLGRVMFKITARA

Biochem/physiol Actions

ATPase Na+/K+ transporting subunit Β 3 (ATP1B3) suppress enterovirus 71 (EV71) replication by elevating the production of type-I interferons, which might act as a potential target in treatment of EV71 infection. The encoded protein is also involved in the regulation of the immune response. Experimental study suggest that ATP1B3 can control restriction of HIV-1 production and nuclear factor Κ B (NF-ΚB) activation in a bone marrow stromal cell antigen 2 (BST-2) dependent manner. Elevated expression of ATP1B3 has been observed in colon and lung cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATP1B3: a virus-induced host factor against EV71 replication by up-regulating the production of type-I interferons.
Lu Y
Virology, 496, 28-34 (2016)
Genetic markers associated with early cancer-specific mortality following prostatectomy.
Liu W
Cancer, 119, 2405-2412 (2013)
Identification and characterization of angiogenesis targets through proteomic profiling of endothelial cells in human cancer tissues.
Mesri M
PLoS ONE, 8 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service