Skip to Content
Merck
All Photos(2)

Documents

SAB1410262

Sigma-Aldrich

Anti-CTNND1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAS, CTNND, KIAA0384, P120CAS, P120CTN, p120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67.1 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNND1(1500)

General description

This gene encodes a member of the Armadillo protein family, which function in adhesion between cells and signal transduction. Multiple translation initiation codons and althernative splicing result in many different isoforms being translated. Not all of the full-length natures of the described transcript variants have been determined. (provided by RefSeq)

Immunogen

CTNND1 (AAH10501.1, 1 a.a. ~ 610 a.a) full-length human protein.

Sequence
MIGEEVPSDQYYWAPLAQHERGSLASLDSLRKGGPPPPNWRQPELPEVIAMLGFRLDAVKSNAAAYLQHLCYRNDKVKTDVRKLKGIPVLVGLLDHPKKEVHLGACGALKNISFGRDQDNKIAIKNCDGVPALVRLLRKARDMDLTEVITGTLWNLSSHDSIKMEIVDHALHALTDEVIIPHSGWEREPNEDCKPRHIEWESVLTNTAGCLRNVSSERSEARRKLRECDGLVDALIFIVQAEIGQKDSDSKLVENCVCLLRNLSYQVHREIPQAERYQEAAPNVANNTGPHAASCFGAKKGKGKKPIEDPANDTVDFPKRTSPARGYELLFQPEVVRIYISLLKESKTPAILEASAGAIQNLCAGRWTYGRYIRSALRQEKALSAIADLLTNEHERVVKAASGALRNLAVDARNKELIGKHAIPNLVKNLPGGQQNSSWNFSEDTVISILNTINEVIAENLEAAKKLRETQGIEKLVLINKSGNRSEKEVRAAALVLQTIWGYKELRKPLEKEGWKKSDFQVNLNNASRSQSSHSYDDSTLPLIDRNQKSDKKPDREEIQMSNMGSNTKSLDNNYSTPNERGDHNKTLDRSGDLGDMEPLKGTTPLMQKI

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Nian et al.
Journal of gynecologic oncology, 34(1), e11-e11 (2022-11-26)
Endometrial carcinoma (EC) is one of the most common malignant tumors of the female reproductive tract, involving multiple molecular alterations. Circular RNA (circRNA) dysregulation is frequently observed in EC tissues, suggesting the involvement of circRNA in EC development. We aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service