Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409477

Sigma-Aldrich

Anti-TRMT12 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ20772, TRM12, TYW2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRMT12(55039)

General description

Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TRMT12 is the human homolog of a yeast gene essential for yW synthesis (Noma and Suzuki, 2006).[supplied by OMIM

Immunogen

TRMT12 (AAH11713.1, 1 a.a. ~ 448 a.a) full-length human protein.

Sequence
MRENVVVSNMERESGKPVAVVAVVTEPRFTQRYREYLQRQKLFDTQHRVEKMPDGSVALPVLGETLPEQHLQELRNRVAPGSPCMLTQLPDPVPSKRAQGCSPAQKLCLEVSRWVEGRGVKWSAELEADLPRSWQRHGNLLLLSEDCFQAKQWKNLGPELWETVALALGVQRLAKRGRVSPDGTRTPAVTLLLGDHGWVEHVDNGIRYKFDVTQCMFSFGNITEKLRVASLSCAGEVLVDLYAGIGYFTLPFLVHAGAAFVHACEWNPHAVVALRNNLEINGVADRCQIHFGDNRKLKLSNIADRVILGLIPSSEEGWPIACQVLRQDAGGILHIHQNVESFPGKNLQALGVSKVEKEHWLYPQQITTNQWKNGATRDSRGKMLSPATKPEWQRWAESAETRIATLLQQVHGKPWKTQILHIQPVKSYAPHVDHIVLDLECCPCPSVG

Biochem/physiol Actions

TRMT12 (tRNA methyltransferase 12 homolog) gene is amplified and over-expressed in breast cancer. This enzyme is responsible for catalyzing the third step in wybutosine biosynthetic pathway. In cancer, up-regulation of this gene leads to disruption in SMG1 protein function, which is a part of nonsense mediated decay pathway (NMD). Thus, it might contribute to the escape of damaged DNA from checkpoints.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosome 8 BAC array comparative genomic hybridization and expression analysis identify amplification and overexpression of TRMT12 in breast cancer.
Rodriguez V et al
Genes Chromosomes Cancer, 46(7), 694-707 (2007)
TRMT12 overexpression in cancer cells affects the cell cycle and the nonsense mediated decay pathways by regulating the SMG-1 kinase, a member of the PIKK family of kinases.
Rodriguez V et al
Cancer Research, 69(9), 2486-2486 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service