Skip to Content
Merck
All Photos(2)

Documents

SAB1408140

Sigma-Aldrich

Anti-TNS4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CTEN, FLJ14950, PP14434

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~51.8 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNS4(84951)

General description

Mouse polyclonal antibody raised against a full-length human TNS4 protein.
TNS4 (Tensin 4) is a signaling molecule localized in the prostate and placenta. It is a member of the TENSIN gene family. It has a Src homology 2 and phosphotyrosine-binding domains. Its expression has been observed in the early progression of melanoma cells.

Immunogen

TNS4 (AAH18706.1, 1 a.a. ~ 480 a.a) full-length human protein.

Sequence
MGSKASSPHGLGSPLVASPRLEKRLGGLAPQRGSRISVLSASPVSDVSYMFGSSQSLLHSSNSSHQSSSRSLESPANSSSSLHSLGSVSLCTRPSDFQAPRNPTLTMGQPRTPHSPPLAKEHASSCPPSITNSMVDIPIVLINGCPEPGSSPPQRTPGHQNSVQPGAASPSNPCPATRSNSQTLSDAPFTTCPEGPARDMQPTMKFVMDTSKYWFKPNITREQAIELLRKEEPGAFVIRDSSSYRGSFGLALKVQEVPASAQSRPGEDSNDLIRHFLIESSAKGVHLKGADEEPYFGSLSAFVCQHSIMALALPCKLTIPQRELGGADGASDSTDSPASCQKKSAGCHTLYLSSVSVETLTGALAVQKAISTTFERDILPTPTVVHFKVTEQGITLTDVQRKVFFRRHYPLTTLRFCGMDPEQRKWQKYCKPSWIFGFVAKSQTEPQENVCHLFAEYDMVQPASQVIGLVTALLQDAERM

Application

Anti-TNS4 antibody produced in mouse is suitable for indirect immunofluorescence and western blot.

Biochem/physiol Actions

TNS4 (Tensin 4) is mainly involved in cell migration activity. It exhibits oncogenic property in colonic, breast, lung, and gastric cancers. In studies, its overexpression has been reported in the pancreatic cancer cells. In pancreatic cancer cells, TNS4 promotes colony formation and cell motility. It has been predicted that TNS4 could be a prognostic marker for primary melanoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Sjoestroem et al.
PloS one, 8(11), e80492-e80492 (2013-11-19)
C-terminal tensin-like protein (Cten) is a focal adhesion protein originally identified as a tumor suppressor in prostate cancer. It has since been found to be overexpressed and function as an oncogene in numerous other cancers, but the expression status of
Saleh Al-Ghamdi et al.
Pancreas, 42(1), 135-140 (2012-07-04)
C-terminal tensin-like gene (CTEN, also known as TNS4) localizes to focal adhesions and is reported to function as an oncogene in colonic, breast, lung, and gastric cancers. Its role in pancreatic cancer is unknown and was thus investigated in this
Abdulkader Albasri et al.
Breast cancer research and treatment, 126(1), 47-54 (2010-04-15)
C-terminal tensin-like (CTEN) gene is a member of the TENSIN gene family, involved in cell migration and localised at focal adhesion sites. This study was designed to explore the prevalence and clinical significance of CTEN expression in a large and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service