Skip to Content
Merck
All Photos(3)

Key Documents

SAB1407815

Sigma-Aldrich

Anti-KIAA1967 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DBC-1, DBC1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~102.9 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

KIAA1967 is a large multi-domain containing nuclear protein mapped on chromosomal location 8p22. It consists of a NL (nuclear localization) motif, a CC (coiled-coil) domain responsible for the protein-protein interaction, an inactive EF-hand for Ca2+ binding, a Nudix domain, a RNA-binding domain and an LZ (leucine zipper) motif.
Mouse polyclonal antibody raised against a full-length human KIAA1967 protein.

Immunogen

KIAA1967 (NP_066997.3, 1 a.a. ~ 923 a.a) full-length human protein.

Sequence
MSQFKRQRINPLPGGRNFSGTASTSLLGPPPGLLTPPVATELSQNARHLQGGEKQRVFTGIVTSLHDYFGVVDEEVFFQLSVVKGRLPQLGEKVLVKAAYNPGQAVPWNAVKVQTLSNQPLLKSPAPPLLHVAALGQKQGILGAQPQLIFQPHRIPPLFPQKPLSLFQTSHTLHLSHLNRFPARGPHGRLDQGRSDDYDSKKRKQRAGGEPWGAKKPRHDLPPYRVHLTPYTVDSPICDFLELQRRYRSLLVPSDFLSVHLSWLSAFPLSQPFSLHHPSRIQVSSEKEAAPDAGAEPITADSDPAYSSKVLLLSSPGLEELYRCCMLFVDDMAEPRETPEHPLKQIKFLLGRKEEEAVLVGGEWSPSLDGLDPQADPQVLVRTAIRCAQAQTGIDLSGCTKWWRFAEFQYLQPGPPRRLQTVVVYLPDVWTIMPTLEEWEALCQQKAAEAAPPTQEAQGETEPTEQAPDALEQAADTSRRNAETPEATTQQETDTDLPEAPPPPLEPAVIARPGCVNLSLHGIVEDRRPKERISFEVMVLAELFLEMLQRDFGYRVYKMLLSLPEKVVSPPEPEKEEAAKEEATKEEEAIKEEVVKEPKDEAQNEGPATESEAPLKEDGLLPKPLSSGGEEEEKPRGEASEDLCEMALDPELLLLRDDGEEEFAGAKLEDSEVRSVASNQSEMEFSSLQDMPKELDPSAVLPLDCLLAFVFFDANWCGYLHRRDLERILLTLGIRLSAEQAKQLVSRVVTQNICQYRSLQYSRQEGLDGGLPEEVLFGNLDLLPPPGKSTKPGAAPTEHKALVSHNGSLINVGSLLQRAEQQDSGRLYLENKIHTLELKLEESHNRFSATEVTNKTLAAEMQELRVRLAEAEETARTAERQKSQLQRLLQELRRRLTPLQLEIQRVVEKADSWVEKEEPAPSN

Biochem/physiol Actions

KIAA1967 is associated with several cellular processes such as regulation of metabolism, aging and tumor biology. It plays a vital role in the cancer pathogenesis by participating in apoptosis, nuclear receptor function, cellular metabolism, circadian cycle and epigenetics. KIAA1967 acts as a regulator of various nuclear receptors such as ERα (oestrogen receptor α), ERβ, AR (androgen receptor), RAR (retinoic acid receptor) and haem receptor Rev-erbα. In addition to transcriptional activity, it also controls epigenetic modifiers such as the deacetylases HDAC3 and SIRT1, and the methyl-transferase SUV39H1. It has been reported that Nudix domain has the ability to bind nucleotides, which further could control its functional activities. In response to UV-induced DNA damage, it plays an important role in the maintenance of genomic stability and cellular integrity. KIAA1967 also possesses a significant role in adipocyte differentiation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José María Moreno-Navarrete et al.
American journal of physiology. Endocrinology and metabolism, 308(7), E554-E561 (2015-02-05)
Genetic deletion of Dbc1 in mice reduced adipose tissue senescence and inflammation while promoting an expansion of this tissue. Here, we aimed to investigate DBC1 mRNA and protein levels in human adipose tissue from subjects with a wide spectrum of
Eduardo Nunes Chini et al.
Bioscience reports, 33(4), doi:10-doi:10 (2013-07-12)
DBC1 (deleted in breast cancer-1) is a nuclear protein that regulates cellular metabolism. Since alteration in cellular metabolism have been proposed to be the emerging 'hallmark' of cancer, it is possible that DBC1 may be implicated in the regulation of
Wootae Kim et al.
Cancer letters, 333(2), 180-186 (2013-01-29)
DBC1 (deleted in breast cancer 1) participates in the regulation of cell survival and death in response to various stimuli. In particular, DBC1 promotes cell death upon DNA damage through inhibition of SIRT1 deacetylase. However, the SIRT1-independent functions of DBC1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service